Learn More
4-Methylthiazole-5-sulfonyl chloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H3373203
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Chemical Identifiers
| 953070-51-4 | |
| 197.651 | |
| AWTJUYXVSWZCIK-UHFFFAOYSA-N | |
| 18537519 | |
| CC1=C(SC=N1)S(=O)(=O)Cl |
| C4H4ClNO2S2 | |
| MFCD09864968 | |
| 4-methylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl, chloro 4-methyl 1,3-thiazol-5-yl sulfone | |
| 4-methyl-1,3-thiazole-5-sulfonyl chloride |
Specifications
| 4-Methylthiazole-5-sulfonyl chloride | |
| C4H4ClNO2S2 | |
| MFCD09864968 | |
| Moisture sensitive | |
| Sparingly soluble in water.(0.26g/L) (25°C), | |
| CC1=C(SC=N1)S(=O)(=O)Cl | |
| 197.651 | |
| 197.66 |
| 953070-51-4 | |
| 1g | |
| UN3261 | |
| 4-methylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl, chloro 4-methyl 1,3-thiazol-5-yl sulfone | |
| AWTJUYXVSWZCIK-UHFFFAOYSA-N | |
| 4-methyl-1,3-thiazole-5-sulfonyl chloride | |
| 18537519 | |
| 95% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
Promotions