accessibility menu, dialog, popup

UserName

CH3SNSOOCl

CAS RN 953070-51-4

IUPAC Name: 4-methyl-1,3-thiazole-5-sulfonyl chloride
Synonyms: 4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 197.65
Molecular Formula: C4H4ClNO2S2
InChi Key: AWTJUYXVSWZCIK-UHFFFAOYSA-N
SMILES: CC1=C(SC=N1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)