missing translation for 'onlineSavingsMsg'
Learn More

4-Methylstyrene (stabilized with TBC) 96.0+%, TCI America™

Catalog No. p-7132164
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
M042825ML 25 mL
M0428100ML 100 mL
M0428500ML 500 mL
3 options

Catalog No. M042825ML

Supplier: TCI America M042825ML

Only null left

Chemical Identifiers

CAS 622-97-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008621
InChI Key JLBJTVDPSNHSKJ-UHFFFAOYSA-N
Synonym 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
PubChem CID 12161
IUPAC Name 1-ethenyl-4-methylbenzene
SMILES CC1=CC=C(C=C)C=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 2618
Formula Weight 118.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylstyrene (stabilized with TBC)
EINECSNumber (3)-0008
RTECSNumber WL5076000
TSCA Yes
Recommended Storage Refrigerator