missing translation for 'onlineSavingsMsg'
Learn More

4-Methylstyrene, 98%, stab. with 0.1% 3,5-di-tert-butylcatechol

CAS: 622-97-9 | C9H10 | 118.18 g/mol

Supplier:  Thermo Scientific Chemicals L0148009

 View more versions of this product

Catalog No. AAL0148009

Only null left
Explore available promotions

Description

Description

4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

Solubility
Miscible with benzene. slightly miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

4-Methylstyrene
622-97-9
0.896
45°C (113°F)
1.543
10 g
1209317
Miscible with benzene. slightly miscible with water.
CC1=CC=C(C=C)C=C1
118.18
118.18
stabilized with 0.1% 3,5-di-tert-butylcatechol
-34°C
171°C to 175°C
C9H10
MFCD00008621
UN2618
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
1-ethenyl-4-methylbenzene
12161
≥98%

Chemical Identifiers

622-97-9
118.18
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
12161
CC1=CC=C(C=C)C=C1
C9H10
MFCD00008621
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
1-ethenyl-4-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319-H335-H340

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED

EINECSNumber : 210-762-8

RTECSNumber : WL5076000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only