missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylpyrimidine 96.0+%, TCI America™
Supplier: TCI America M035625G
Specifications
| 4-Methylpyrimidine | |
| Yellow | |
| C5H6N2 | |
| 25 g | |
| pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci | |
| CC1=NC=NC=C1 | |
| 94.117 | |
| 94.12 | |
| Liquid |
| 3438-46-8 | |
| 142°C | |
| MFCD00006115 | |
| 1993 | |
| LVILGAOSPDLNRM-UHFFFAOYSA-N | |
| 4-methylpyrimidine | |
| 18922 | |
| ≥96.0% (T) |
Chemical Identifiers
| 3438-46-8 | |
| 94.117 | |
| LVILGAOSPDLNRM-UHFFFAOYSA-N | |
| 18922 | |
| CC1=NC=NC=C1 |
| C5H6N2 | |
| MFCD00006115 | |
| pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci | |
| 4-methylpyrimidine |
Safety and Handling
TSCA : Yes