accessibility menu, dialog, popup

UserName

CH3NN

CAS RN 3438-46-8

IUPAC Name: 4-methylpyrimidine
Synonyms: 4-methylpyrimidine
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: LVILGAOSPDLNRM-UHFFFAOYSA-N
SMILES: CC1=CC=NC=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)