missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyrimidine 96.0+%, TCI America™

Catalog No. p-7132118
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03565G 5 g
M035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03565G Supplier TCI America Supplier No. M03565G
Only null left

Chemical Identifiers

CAS 3438-46-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006115
InChI Key LVILGAOSPDLNRM-UHFFFAOYSA-N
Synonym pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci
PubChem CID 18922
IUPAC Name 4-methylpyrimidine
SMILES CC1=NC=NC=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 1993
Formula Weight 94.12
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Methylpyrimidine