missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphthalic Acid 98.0+%, TCI America™

Catalog No. M056025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M056025G 25 g
M0560500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M056025G Supplier TCI America Supplier No. M056025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4316-23-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00041946
InChI Key CWJJAFQCTXFSTA-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-methyl, phthalic acid, 4-methyl, 4-methyl phthalic acid, 4-methyl-phthalic acid, 4-methylbenzene-1,2-dicarboxylic acid, 4-methyl-1,2-benzenedicarboxylic acid, 4-methyl-o-phthalate, 4-methylphthalicacid, acmc-1amca, 4-methylphthalic acid
PubChem CID 20310
IUPAC Name 4-methylphthalic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)C(=O)O

Specifications

Melting Point 147°C
pH 5
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylphthalic Acid
EINECSNumber (3)-1373
TSCA Yes