accessibility menu, dialog, popup

UserName

CH3OHOHOO

CAS RN 4316-23-8

IUPAC Name: 4-methylbenzene-1,2-dicarboxylic acid
Synonyms: 4-methylbenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: CWJJAFQCTXFSTA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=O)C(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results