missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenylacetone, 97+%

Catalog No. p-7027737
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2472803 1 g
AAB2472806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2472803 Supplier Thermo Scientific Chemicals Supplier No. B2472803

CAS: 2096-86-8 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2096-86-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00017250
InChI Key NOXKUHSBIXPZBJ-UHFFFAOYSA-N
Synonym 4-methylphenylacetone, 1-4-methylphenyl propan-2-one, 1-p-tolyl propan-2-one, 4-methylphenyl acetone, 1-4-methylphenyl acetone, p-methylphenyl acetone, 1-4-methylphenyl-2-propanone, 2-propanone,1-4-methylphenyl, p-methylphenylacetone, acmc-20amd6
PubChem CID 137428
IUPAC Name 1-(4-methylphenyl)propan-2-one
SMILES CC1=CC=C(C=C1)CC(=O)C

Specifications

Quantity 1 g
Formula Weight 148.2
Percent Purity 98%
Chemical Name or Material 4-Methylphenylacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only