accessibility menu, dialog, popup

UserName

CAS RN 2096-86-8

IUPAC Name: 1-(4-methylphenyl)propan-2-one
Synonyms: 1-(4-methylphenyl)propan-2-one
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: NOXKUHSBIXPZBJ-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results