missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™

$221.83 - $641.24

Chemical Identifiers

CAS 1204518-02-4
Molecular Formula C17H18F3IO3S
Molecular Weight (g/mol) 486.288
MDL Number MFCD20264882
InChI Key KVLSSMIQFXWHII-UHFFFAOYSA-M
Synonym Mesityl(p-tolyl)iodonium Triflate
PubChem CID 71311054
IUPAC Name (4-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate
SMILES CC1=CC=C(C=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29091G
View Documents
TCI America
M29091G
1 g N/A
Only null left
 
M29095G
View Documents
TCI America
M29095G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1204518-02-4
486.288
KVLSSMIQFXWHII-UHFFFAOYSA-M
71311054
CC1=CC=C(C=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
C17H18F3IO3S
MFCD20264882
Mesityl(p-tolyl)iodonium Triflate
(4-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate

Specifications

1204518-02-4
C17H18F3IO3S
1 g
KVLSSMIQFXWHII-UHFFFAOYSA-M
(4-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate
71311054
≥98.0% (HPLC,T)
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate
179°C
MFCD20264882
Mesityl(p-tolyl)iodonium Triflate
CC1=CC=C(C=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]
486.288
486.29
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'