CAS RN 1204518-02-4
CAS RN 1204518-02-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
CAS: 1204518-02-4 Molecular Formula: C17H18F3IO3S Molecular Weight (g/mol): 486.288 MDL Number: MFCD20264882 InChI Key: KVLSSMIQFXWHII-UHFFFAOYSA-M Synonym: Mesityl(p-tolyl)iodonium Triflate PubChem CID: 71311054 IUPAC Name: (4-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate SMILES: CC1=CC=C(C=C1)[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]