missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcatechol 98.0+%, TCI America™

Catalog No. p-7132153
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M041325G 25 g
M0413500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M041325G Supplier TCI America Supplier No. M041325G
Only null left

Chemical Identifiers

CAS 452-86-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002205
InChI Key ZBCATMYQYDCTIZ-UHFFFAOYSA-N
Synonym 4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
PubChem CID 9958
ChEBI CHEBI:17254
IUPAC Name 4-methylbenzene-1,2-diol
SMILES CC1=CC=C(O)C(O)=C1

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 117°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylcatechol
EINECSNumber (4)-0057
RTECSNumber UX1915000
TSCA No