missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcatechol, 96%

CAS: 452-86-8 | C7H8O2 | 124.14 g/mol

Supplier:  Thermo Scientific Chemicals L0449536

 View more versions of this product

Catalog No. AAL0449536


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Methylcatechol is used to synthesize antimicrobial, antioxidant. It also acts as effective inhibitor. It can be used to produce 4-bromo-5-methyl-pyrocatechol. It Increases Brain-Derived Neurotrophic Factor Content and mRNA Expression in Cultured Brain Cells.

Solubility
Soluble in water, methanol, acetone, ether, and alcohol.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

4-Methylcatechol
67°C to 69°C
140°C (284°F)
MFCD00002205
636512
Soluble in water,methanol,acetone,ether,and alcohol.
CC1=CC=C(O)C(O)=C1
124.14
CHEBI:17254
96%
452-86-8
251°C
C7H8O2
500 g
4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
ZBCATMYQYDCTIZ-UHFFFAOYSA-N
4-methylbenzene-1,2-diol
9958
124.14

Chemical Identifiers

452-86-8
124.14
ZBCATMYQYDCTIZ-UHFFFAOYSA-N
9958
4-methylbenzene-1,2-diol
C7H8O2
MFCD00002205
4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
CHEBI:17254
CC1=CC=C(O)C(O)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

EINECSNumber : 207-214-5

RTECSNumber : UX1915000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only