missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. AC450410010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450410010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450410010 Supplier Thermo Scientific Chemicals Supplier No. 450410010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101366-51-2
Molecular Formula C13H11ClO2S
Molecular Weight (g/mol) 266.75
InChI Key DBYXACYHGMSPMQ-UHFFFAOYSA-N
Synonym 4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
PubChem CID 2794758
IUPAC Name 4-(4-methylphenyl)benzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1g
Formula Weight 266.75
Percent Purity 97%
Chemical Name or Material 4'-Methylbiphenyl-4-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.

RUO – Research Use Only