CAS RN 101366-51-2
IUPAC Name:
4'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms:
4'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol):
266.74
Molecular Formula:
C13H11ClO2S
InChi Key:
DBYXACYHGMSPMQ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
Promotions Available
4'-Methylbiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™
4'-Methylbiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™
CAS: 101366-51-2 Molecular Formula: C13H11ClO2S Molecular Weight (g/mol): 266.75 InChI Key: DBYXACYHGMSPMQ-UHFFFAOYSA-N Synonym: 4'-methylbiphenyl-4-sulfonyl chloride,4'-methyl 1,1'-biphenyl-4-sulfonyl chloride,4-4-methylphenyl benzenesulfonyl chloride,4-4-methylphenyl phenyl sulphonyl chloride,4'-methyl-1,1'-biphenyl-4-sulfonyl chloride,4-p-tolyl benzenesulfonyl chloride,acmc-20amef,4-4-chlorosulphonyl phenyl toluene,4-chlorosulphonyl-4'-methylbiphenyl PubChem CID: 2794758 IUPAC Name: 4-(4-methylphenyl)benzenesulfonyl chloride SMILES: CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl