missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-carboxylic acid, 96%

Catalog No. AAH3445906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3445906 5 g
AAH3445903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3445906 Supplier Thermo Scientific Chemicals Supplier No. H3445906

CAS: 720-73-0 | C14H12O2 | 212.248 g/mol

The 4'-methyl-biphenyl-4-carboxylic acid exhibits the characteristic absorption bands of COOH and CH, groups. Photoproduct analysis, carried out by GC-MS, ETIR and GC-FTIR confirm three successive oxidation states of methyl suhstituents such as alcohol, aldehyde and carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The 4′-methyl-biphenyl-4-carboxylic acid exhibits the characteristic absorption bands of COOH and CH, groups. Photoproduct analysis, carried out by GC-MS, ETIR and GC-FTIR confirm three successive oxidation states of methyl suhstituents such as alcohol, aldehyde and carboxylic acids.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 720-73-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00448778
InChI Key RZOCCLOTCXINRG-UHFFFAOYSA-N
Synonym 4'-methylbiphenyl-4-carboxylic acid, 4-4-methylphenyl benzoic acid, 4'-methyl-1,1'-biphenyl-4-carboxylic acid, 4'-methyl 1,1'-biphenyl-4-carboxylic acid, 4'-methyl-4-biphenylcarboxylic acid, 4'-methyl-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylicacid, 4'-methyl, 1,1'-biphenyl-4-carboxylic acid, 4'-methyl
PubChem CID 2063421
IUPAC Name 4-(4-methylphenyl)benzoic acid
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 257°C to 258°C
Quantity 5 g
Formula Weight 212.25
Percent Purity 96%
Chemical Name or Material 4'-Methylbiphenyl-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only