accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 720-73-0

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-4-carboxylate
Synonyms: 4'-methyl-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 211.24
Molecular Formula: C14H11O2
InChi Key: RZOCCLOTCXINRG-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)C1=CC=C(C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results