missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine, 98%

Catalog No. p-159718
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126400050 5 g Glass bottle
AC126400250 25 g Glass bottle
AC126401000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126400050 Supplier Thermo Scientific Chemicals Supplier No. 126400050
Only null left

CAS: 104-84-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-84-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008123
InChI Key HMTSWYPNXFHGEP-UHFFFAOYSA-N
Synonym 4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
PubChem CID 66035
IUPAC Name (4-methylphenyl)methanamine
SMILES CC1=CC=C(C=C1)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 13°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 195°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.536
Linear Formula CH3C6H4CH2NH2
Quantity 5 g
Beilstein 12, 1141
Specific Gravity 0.95
Solubility Information Solubility in water: slightly soluble
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 203-243-2
TSCA TSCA

RUO – Research Use Only