missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl isothiocyanate, 96%

Catalog No. AAL1113504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1113504 2 g
AAL1113509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1113504 Supplier Thermo Scientific Chemicals Supplier No. L1113504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3694-46-0 | C9H9NS | 163.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3694-46-0
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00041118
InChI Key OAXIUBJXQISJEV-UHFFFAOYSA-N
Synonym 4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference
PubChem CID 19409
IUPAC Name 1-(isothiocyanatomethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CN=C=S

Specifications

Density 1.08
Boiling Point 97°C (1 mmHg)
Odor Pungent
Refractive Index 1.589
Quantity 2 g
UN Number UN2922
Beilstein 2082478
Sensitivity Moisture sensitive
Formula Weight 163.24
Percent Purity 96%
Chemical Name or Material 4-Methylbenzyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
RTECSNumber NX8860000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only