missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzotrifluoride 98.0+%, TCI America™

Catalog No. M294525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M294525G 25 g
M29455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M294525G Supplier TCI America Supplier No. M294525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6140-17-6
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.139
MDL Number MFCD00075476
InChI Key LRLRAYMYEXQKID-UHFFFAOYSA-N
Synonym 4-methylbenzotrifluoride, 1-methyl-4-trifluoromethyl benzene, 4-trifluoromethyl toluene, p-methylbenzotrifluoride, alpha,alpha,alpha-trifluoro-p-xylene, benzene, 1-methyl-4-trifluoromethyl, 4-methyl-1-trifluoromethyl benzene, 1-methyl-4-trifluormethyl benzol, pubchem1659, p-trifluoromethyltoluene
PubChem CID 80230
IUPAC Name 1-methyl-4-(trifluoromethyl)benzene
SMILES CC1=CC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 129°C
Quantity 25 g
UN Number 1993
Formula Weight 160.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylbenzotrifluoride