accessibility menu, dialog, popup

UserName

CH3FFF

CAS RN 6140-17-6

IUPAC Name: 1-methyl-4-(trifluoromethyl)benzene
Synonyms: 1-methyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 160.14
Molecular Formula: C8H7F3
InChi Key: LRLRAYMYEXQKID-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)