missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzotrifluoride, 98%

Catalog No. AC308910010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308910010 1 g Glass bottle
AC308910050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308910010 Supplier Thermo Scientific Chemicals Supplier No. 308910010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6140-17-6 | C8H7F3 | 160.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6140-17-6
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.14
MDL Number MFCD00075476
InChI Key LRLRAYMYEXQKID-UHFFFAOYSA-N
Synonym 4-methylbenzotrifluoride, 1-methyl-4-trifluoromethyl benzene, 4-trifluoromethyl toluene, p-methylbenzotrifluoride, alpha,alpha,alpha-trifluoro-p-xylene, benzene, 1-methyl-4-trifluoromethyl, 4-methyl-1-trifluoromethyl benzene, 1-methyl-4-trifluormethyl benzol, pubchem1659, p-trifluoromethyltoluene
PubChem CID 80230
IUPAC Name 1-methyl-4-(trifluoromethyl)benzene
SMILES CC1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1440g/mL
Boiling Point 129°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4250 to 1.427
Linear Formula CH3C6H4CF3
Quantity 1 g
Specific Gravity 1.144
Formula Weight 160.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzotrifluoride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-124-2

RUO – Research Use Only