missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. M24235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24235G Supplier TCI America Supplier No. M24235G

Chemical Identifiers

CAS 41263-74-5
Molecular Formula C8H8N2O4
Molecular Weight (g/mol) 196.162
MDL Number MFCD00464322
InChI Key KSMLIIWEQBYUKA-UHFFFAOYSA-N
PubChem CID 283475
SMILES CNC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 310°C
Color Yellow
Quantity 5 g
Formula Weight 196.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylamino)-3-nitrobenzoic Acid