accessibility menu, dialog, popup

UserName

H3CNHNOOOO

CAS RN 41263-74-5

IUPAC Name: 4-(methylamino)-3-nitrobenzoate
Synonyms: 4-(methylamino)-3-nitrobenzoate
Molecular Weight (g/mol): 195.16
Molecular Formula: C8H7N2O4
InChi Key: KSMLIIWEQBYUKA-UHFFFAOYSA-M
SMILES: CNC1=CC=C(C=C1[N+]([O-])=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results