missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-N-tosylbenzenesulfonamide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 173°C |
| Color | White |
| Boiling Point | 265°C |
| Quantity | 5 g |
| Formula Weight | 325.40 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methyl-N-tosylbenzenesulfonamide |
Safety and Handling
| TSCA | Yes |