accessibility menu, dialog, popup

UserName

CAS RN 3695-00-9

IUPAC Name: 4-methyl-N-(4-methylbenzenesulfonyl)benzene-1-sulfonamide
Synonyms: 4-methyl-N-(4-methylbenzenesulfonyl)benzenesulfonamide
Molecular Weight (g/mol): 325.4
Molecular Formula: C14H15NO4S2
InChi Key: LHWZLUXODWUHLZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results