missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. M27085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27085G 5 g
M27081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27085G Supplier TCI America Supplier No. M27085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 261952-01-6
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD01631592
InChI Key CAPKAYDTKWGFQB-UHFFFAOYSA-N
Synonym 4-methyl-3-trifluoromethyl benzoic acid, 3-trifluoromethyl-4-methyl benzoic acid, 3-trifluoromethyl-p-toluic acid, mtf-boa-4me, 4-methyl-3-trifluoromethyl benzoicacid, 4-methyl-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-methylbenzoic acid, benzoic acid, 4-methyl-3-trifluoromethyl, pubchem2701
PubChem CID 2775592
IUPAC Name 4-methyl-3-(trifluoromethyl)benzoic acid
SMILES CC1=CC=C(C=C1C(F)(F)F)C(O)=O

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzoic Acid