accessibility menu, dialog, popup

UserName

CAS RN 261952-01-6

IUPAC Name: 4-methyl-3-(trifluoromethyl)benzoic acid
Synonyms: 4-methyl-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: CAPKAYDTKWGFQB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C(F)(F)F)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results