missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. p-7133354
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26591G 1 g
M26595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26591G Supplier TCI America Supplier No. M26591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65934-74-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD01631582
InChI Key JBCDCYFEJQHTTA-UHFFFAOYSA-N
Synonym 4-methyl-3-trifluoromethyl aniline, 5-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-4-methylaniline, benzenamine, 4-methyl-3-trifluoromethyl, 4-methyl-5-trifluoromethyl aniline, 4-methyl-3-trifluoromethyl-phenylamine, 4-methyl-5-trifluoromethylaniline, 3-trifluoromethyl-p-toluidine, 4-methyl-3-trifluoromethyl phenylamine
PubChem CID 2737715
IUPAC Name 4-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=C(C=C1)N)C(F)(F)F

Specifications

Color Yellow
Boiling Point 204°C
Quantity 1 g
Formula Weight 175.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)aniline