missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzamide, 98%

Catalog No. AAL1827014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1827014 25 g
AAL1827006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1827014 Supplier Thermo Scientific Chemicals Supplier No. L1827014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19013-11-7 | C8H8N2O3 | 180.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19013-11-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00027392
InChI Key YEUGEQUFPMJGCD-UHFFFAOYSA-N
Synonym 3-nitro-4-methylbenzamide, benzamide, 4-methyl-3-nitro, benzamide,4-methyl-3-nitro, acmc-20amdw
PubChem CID 87896
IUPAC Name 4-methyl-3-nitrobenzamide
SMILES CC1=C(C=C(C=C1)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 164°C to 167°C
Quantity 25 g
Beilstein 2102644
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 242-752-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only