accessibility menu, dialog, popup

UserName

CAS RN 19013-11-7

IUPAC Name: 4-methyl-3-nitrobenzamide
Synonyms: 4-methyl-3-nitrobenzamide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: YEUGEQUFPMJGCD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results