missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzaldehyde, 98%

Catalog No. AAL1345406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1345406 5 g
AAL1345403 1 g
AAL1345414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1345406 Supplier Thermo Scientific Chemicals Supplier No. L1345406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31680-07-6 | C8H7NO3 | 165.15 g/mol

4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 31680-07-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00017011
InChI Key KHWGAWBXQOKXIJ-UHFFFAOYSA-N
Synonym 3-nitro-p-tolualdehyde, 4-methyl-3-nitro-benzaldehyde, 3-nitro-4-methylbenzaldehyde, benzaldehyde, 4-methyl-3-nitro, methylnitrobenzaldehyde, pubchem8157, acmc-209hod, 4-methyl-3-nitrobenzaldehyde
PubChem CID 1798998
IUPAC Name 4-methyl-3-nitrobenzaldehyde
SMILES CC1=CC=C(C=O)C=C1[N+]([O-])=O

Specifications

Melting Point 44°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1637706
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-760-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only