CAS RN 31680-07-6
IUPAC Name:
4-methyl-3-nitrobenzaldehyde
Synonyms:
4-methyl-3-nitrobenzaldehyde
Molecular Weight (g/mol):
165.15
Molecular Formula:
C8H7NO3
InChi Key:
KHWGAWBXQOKXIJ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=O)C=C1[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
4-Methyl-3-nitrobenzaldehyde, 98%
4-Methyl-3-nitrobenzaldehyde, 98%
CAS: 31680-07-6 Molecular Formula: C8H7NO3 Molecular Weight (g/mol): 165.15 MDL Number: MFCD00017011 InChI Key: KHWGAWBXQOKXIJ-UHFFFAOYSA-N Synonym: 3-nitro-p-tolualdehyde,4-methyl-3-nitro-benzaldehyde,3-nitro-4-methylbenzaldehyde,benzaldehyde, 4-methyl-3-nitro,methylnitrobenzaldehyde,pubchem8157,acmc-209hod,4-methyl-3-nitrobenzaldehyde PubChem CID: 1798998 IUPAC Name: 4-methyl-3-nitrobenzaldehyde SMILES: CC1=CC=C(C=O)C=C1[N+]([O-])=O