missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. M18825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18825G 5 g
M18821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18825G Supplier TCI America Supplier No. M18825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 262373-15-9
Molecular Formula C11H15F3O3SSi
Molecular Weight (g/mol) 312.378
InChI Key SKLVYEYSGSTOFD-UHFFFAOYSA-N
Synonym 4-Methyl-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulfonate
PubChem CID 11449792
IUPAC Name (4-methyl-2-trimethylsilylphenyl) trifluoromethanesulfonate
SMILES CC1=CC(=C(C=C1)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 312.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate