accessibility menu, dialog, popup

UserName

CH3OSOOFFFSiCH3H3CCH3

CAS RN 262373-15-9

IUPAC Name: 4-methyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms: 4-methyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 312.38
Molecular Formula: C11H15F3O3SSi
InChi Key: SKLVYEYSGSTOFD-UHFFFAOYSA-N
SMILES: CC1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results