missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Methyl-2-pentanol, 99%

CAS: 108-11-2 | C6H14O | 102.177 g/mol

Supplier:  Thermo Scientific Chemicals A13435AP

 View more versions of this product

Catalog No. AAA13435AP


Only null left
Explore available promotions

Description

Description

(±)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.

Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and acid chlorides.
Specifications

Specifications

(+/-)-4-Methyl-2-pentanol
-90°C
131°C to 133°C
Slightly Sweetish
1.411
500 mL
1098268
Miscible with ethanol and diethyl ether. Slightly miscible with water.
CC(C)CC(C)O
102.177
102.18
108-11-2
0.806
41°C (105°F)
C6H14O
MFCD00004550
UN2053
4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol
WVYWICLMDOOCFB-UHFFFAOYSA-N
4-methylpentan-2-ol
7910
99%

Chemical Identifiers

108-11-2
102.177
WVYWICLMDOOCFB-UHFFFAOYSA-N
7910
CC(C)CC(C)O
C6H14O
MFCD00004550
4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol
4-methylpentan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H335
Flammable liquid and vapour.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL ISOBUTYL CARBINOL

EINECSNumber : 203-551-7

RTECSNumber : SA7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only