Learn More
(+/-)-4-Methyl-2-pentanol, 99%
Description
(±)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(^+)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.
Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and acid chlorides.
Specifications
Specifications
| Melting Point | -90°C |
| Density | 0.806 |
| Boiling Point | 131°C to 133°C |
| Flash Point | 41°C (105°F) |
| Odor | Slightly Sweetish |
| Refractive Index | 1.411 |
| Quantity | 500 mL |
| UN Number | UN2053 |
| Beilstein | 1098268 |
| Solubility Information | Miscible with ethanol and diethyl ether. Slightly miscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H335 |
| Hazard Statement | GHS H Statement H226-H335 Flammable liquid and vapour. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL ISOBUTYL CARBINOL |
| EINECSNumber | 203-551-7 |
| RTECSNumber | SA7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only