accessibility menu, dialog, popup

UserName

CAS RN 108-11-2

IUPAC Name: 4-methylpentan-2-ol
Synonyms: 4-methyl-2-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: WVYWICLMDOOCFB-UHFFFAOYNA-N
SMILES: CC(C)CC(C)O
Boiling Point 
  • (2)
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)