missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenol, 97%

Catalog No. AAA1200914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1200914 25 g
AAA1200922 100 g
AAA1200936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1200914 Supplier Thermo Scientific Chemicals Supplier No. A1200914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119-33-5 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 32°C to 35°C
Density 1.24
Boiling Point 125°C (22 mmHg)
Flash Point 108°C (226°F)
Odor Phenol-like
Quantity 25 g
Beilstein 1868022
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-nitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-315-6
RTECSNumber GP2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only