accessibility menu, dialog, popup

UserName

CAS RN 119-33-5

IUPAC Name: 4-methyl-2-nitrophenol
Synonyms: 2-nitro-P-cresol nitrocresols and their alkali metal salts
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: SYDNSSSQVSOXTN-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=C1)[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)