missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals

Catalog No. AAH2879506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2879506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2879506 Supplier Thermo Scientific Chemicals Supplier No. H2879506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14001-63-9 | Molecular Formula: C6H8N2S | Molecular Weight: 140.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14001-63-9
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.204
MDL Number MFCD00023242
InChI Key UCERVHYBSTYCQS-UHFFFAOYSA-N
Synonym 4-methyl-2-methylthio pyrimidine, 4-methyl-2 methylsulfanyl pyrimidine, 4-methyl-2-methylsulfanyl pyrimidine, pyrimidine, 4-methyl-2-methylthio, 4-methyl-2-methylsulfanyl-pyrimidine, 2-methylthio-4-methylpyrimidine, 4-methyl-2-methylthiopyrimidine, 4-methyl-2-methylsulphanyl pyrimidine, acmc-1bust, methylmethylsulfanylpyrimidine
PubChem CID 821261
IUPAC Name 4-methyl-2-methylsulfanylpyrimidine
SMILES CC1=NC(=NC=C1)SC

Specifications

Density 1.01
Flash Point 104°C (219°F)
Refractive Index 1.571
Quantity 5 g
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(methylthio)pyrimidine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only