accessibility menu, dialog, popup

UserName

CAS RN 14001-63-9

IUPAC Name: 4-methyl-2-(methylsulfanyl)pyrimidine
Synonyms: 4-methyl-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 140.2
Molecular Formula: C6H8N2S
InChi Key: UCERVHYBSTYCQS-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results