missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxytrityl alcohol, 94%

CAS: 847-83-6 | C20H18O2 | 290.36 g/mol

$95.68 - $1,355.75

Chemical Identifiers

CAS 847-83-6
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.36
MDL Number MFCD00087962
InChI Key WCRRRAKYYPJJMP-UHFFFAOYSA-N
Synonym p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole
PubChem CID 70061
IUPAC Name (4-methoxyphenyl)-diphenylmethanol
SMILES COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1862009
View Documents
Thermo Scientific Chemicals
A1862009
10 g N/A
Only null left
 
AAA1862018
View Documents
Thermo Scientific Chemicals
A1862018
50 g N/A
Only null left
 
AAA1862030
View Documents
Thermo Scientific Chemicals
A1862030
250 g N/A
Only null left
 
Description

Description

It is used as an intermediate for preparation of TT-0011.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate for preparation of TT-0011.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

847-83-6
290.36
WCRRRAKYYPJJMP-UHFFFAOYSA-N
70061
COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
C20H18O2
MFCD00087962
p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole
(4-methoxyphenyl)-diphenylmethanol

Specifications

847-83-6
MFCD00087962
p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole
WCRRRAKYYPJJMP-UHFFFAOYSA-N
(4-methoxyphenyl)-diphenylmethanol
70061
94%
C20H18O2
10 g
Insoluble in water.
COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
290.36
290.36
4-Methoxytrityl alcohol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-693-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only