Learn More
4-Methoxytrityl alcohol, 94%
CAS: 847-83-6 | C20H18O2 | 290.36 g/mol
Supplier: Thermo Scientific Chemicals A1862030
Description
It is used as an intermediate for preparation of TT-0011.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an intermediate for preparation of TT-0011.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-Methoxytrityl alcohol | |
| C20H18O2 | |
| 250 g | |
| Insoluble in water. | |
| COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 290.36 | |
| 290.36 |
| 847-83-6 | |
| MFCD00087962 | |
| p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole | |
| WCRRRAKYYPJJMP-UHFFFAOYSA-N | |
| (4-methoxyphenyl)-diphenylmethanol | |
| 70061 | |
| 94% |
Chemical Identifiers
| 847-83-6 | |
| 290.36 | |
| WCRRRAKYYPJJMP-UHFFFAOYSA-N | |
| 70061 | |
| COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H18O2 | |
| MFCD00087962 | |
| p-methoxytrityl alcohol, 4-methoxyphenyl diphenylmethanol, unii-d117s0gbot, 4-methoxytrityl alcohol, p-methoxyphenyl-diphenylmethanol, p-anisyldiphenylmethanol, d117s0gbot, 4-methoxyphenyldiphenylmethanol, methoxytriphenylcarbinole | |
| (4-methoxyphenyl)-diphenylmethanol |
Safety and Handling
EINECSNumber : 212-693-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only