missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystilbene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1732806

 View more versions of this product

Catalog No. AAA1732806

Only null left
Explore available promotions

Description

Description

It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.

Solubility
Sparingly soluble (0.061 g/L) (25°C) in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

4-Methoxystilbene
136°C to 138°C
MFCD00017178
p-methoxystilbene, 4-methoxystilbene, anisole, p-styryl, trans-4-methoxystilbene, e-4-methoxystilbene, 1-methoxy-4-e-2-phenylethenyl benzene, 1-p-methoxyphenyl-2-phenylethene, unii-5el148b7hg, benzene, 1-methoxy-4-2-phenylethenyl, 1-methoxy-4-e-styryl-benzene
XWYXLYCDZKRCAD-BQYQJAHWSA-N
1-methoxy-4-[(E)-2-phenylethenyl]benzene
1245920
210.27
1142-15-0
C15H14O
5 g
Sparingly soluble (0.061g/L) (25°C) in water.
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
210.276
CHEBI:35094
98%

Chemical Identifiers

1142-15-0
210.276
XWYXLYCDZKRCAD-BQYQJAHWSA-N
1245920
1-methoxy-4-[(E)-2-phenylethenyl]benzene
C15H14O
MFCD00017178
p-methoxystilbene, 4-methoxystilbene, anisole, p-styryl, trans-4-methoxystilbene, e-4-methoxystilbene, 1-methoxy-4-e-2-phenylethenyl benzene, 1-p-methoxyphenyl-2-phenylethene, unii-5el148b7hg, benzene, 1-methoxy-4-2-phenylethenyl, 1-methoxy-4-e-styryl-benzene
CHEBI:35094
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 214-530-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only