missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystilbene, 98%, Thermo Scientific Chemicals

Catalog No. p-7023602
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1732806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1732806 Supplier Thermo Scientific Chemicals Supplier No. A1732806

It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.

Solubility
Sparingly soluble (0.061 g/L) (25°C) in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1142-15-0
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017178
InChI Key XWYXLYCDZKRCAD-BQYQJAHWSA-N
Synonym p-methoxystilbene, 4-methoxystilbene, anisole, p-styryl, trans-4-methoxystilbene, e-4-methoxystilbene, 1-methoxy-4-e-2-phenylethenyl benzene, 1-p-methoxyphenyl-2-phenylethene, unii-5el148b7hg, benzene, 1-methoxy-4-2-phenylethenyl, 1-methoxy-4-e-styryl-benzene
PubChem CID 1245920
ChEBI CHEBI:35094
IUPAC Name 1-methoxy-4-[(E)-2-phenylethenyl]benzene
SMILES COC1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 136°C to 138°C
Quantity 5 g
Solubility Information Sparingly soluble (0.061g/L) (25°C) in water.
Formula Weight 210.27
Percent Purity 98%
Chemical Name or Material 4-Methoxystilbene
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 214-530-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only