missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxysalicylamide 98.0+%, TCI America™

Catalog No. M18145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18145G 5 g
M181425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18145G Supplier TCI America Supplier No. M18145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6745-77-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD03840504
InChI Key BCYRZPCPWKCINY-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-methoxybenzamide
PubChem CID 4868377
IUPAC Name 2-hydroxy-4-methoxybenzamide
SMILES COC1=CC=C(C(N)=O)C(O)=C1

Specifications

Melting Point 158°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxysalicylamide