accessibility menu, dialog, popup

UserName

H3CONH2OHO

CAS RN 6745-77-3

IUPAC Name: 2-hydroxy-4-methoxybenzamide
Synonyms: 2-hydroxy-4-methoxybenzamide
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: BCYRZPCPWKCINY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(N)=O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results